4-Bromo-2-iodo-1-(trifluoromethyl)benzene ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Bromo-2-iodobenzotrifluoride

075918-1G 075918-5G 075918-10G
102543-626EA 143.08 USD
102543-626 102543-628 102543-630
4-Bromo-2-iodo-1-(trifluoromethyl)benzene ≥95%
4-Bromo-2-iodo-1-(trifluoromethyl)benzene

Hazard: Irritant

Formula: C₇H₃BrF₃I
MW: 350.91 g/mol
MDL Number: MFCD21603817
CAS Number: 1256945-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR