3-Bromo-5-ethylpyridine ≥95%

Supplier: Matrix Scientific

075916-250MG 075916-1G
102543-616EA 498.46 USD
102543-616 102543-618
3-Bromo-5-ethylpyridine ≥95%
3-Bromo-5-ethylpyridine

Hazard: Irritant

Formula: C7H8BrN
MW: 186.0491 g/mol
MDL Number: MFCD09800592
CAS Number: 142337-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR