1-(2-Aminothiazol-4-yl)ethanone ≥95%

Supplier: Matrix Scientific

075915-250MG 075915-1G
102543-612EA 569.23 USD
102543-612 102543-614
1-(2-Aminothiazol-4-yl)ethanone ≥95%
1-(2-Aminothiazol-4-yl)ethanone

Hazard: Irritant

Formula: C₅H₆N₂OS
MW: 142.1800 g/mol
MDL Number: MFCD06090850
CAS Number: 101258-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR