1-Bromo-3-nitrobenzene ≥95%

Supplier: Matrix Scientific

075914-100G 075914-25G
102543-610EA 93.85 USD
102543-610 102833-888
1-Bromo-3-nitrobenzene ≥95%
1-Bromo-3-nitrobenzene

Hazard: Toxic

Formula: C₆H₄BrNO₂
MW: 202.01 g/mol
Boiling Pt: 256 °C (760 mmHg)
Melting Pt: 51…54 °C
Density: 1.704
Flash Pt: 113 °C
MDL Number: MFCD00024298
CAS Number: 585-79-5
UN: 3459
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR