Methyl-3-bromo-2-fluorobenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-bromo-2-fluorobenzoate

075913-1G 075913-5G
102543-602EA 52.31 USD
102543-602 102543-604
Methyl-3-bromo-2-fluorobenzoate ≥95%
Methyl 3-bromo-2-fluorobenzoate

Hazard: Irritant

Formula: C₈H₆BrFO₂
MDL Number: MFCD09261255
CAS Number: 206551-41-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR