5-Bromo-3-hydroxy-2-picoline ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Bromo-2-methylpyridin-3-ol
075910-1G
075910-5G
102543-586EA
783.08
USD
102543-586
102543-588
5-Bromo-3-hydroxy-2-picoline ≥95%
5-Bromo-3-hydroxy-2-picoline
Hazard: Irritant
Formula:
C₆H₆BrNO MW: 188.0200 g/mol |
MDL Number:
MFCD12024425 CAS Number: 91420-25-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...