7-Methyl-1H-indole-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

075909-1G 075909-5G
102543-582EA 366.15 USD
102543-582 102543-584
7-Methyl-1H-indole-3-carboxylic acid ≥95%
7-Methyl-1H-indole-3-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₉NO₂
MW: 175.1800 g/mol
MDL Number: MFCD06203634
CAS Number: 30448-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR