(R)-Morpholin-3-ylmethanol hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: (3R)-3-(Hydroxymethyl)morpholine hydrochloride

075907-250MG 075907-1G
102543-572EA 323.08 USD
102543-572 102543-574
(R)-Morpholin-3-ylmethanol hydrochloride ≥95%
(R)-Morpholin-3-ylmethanol hydrochloride

Hazard: Irritant-Harmful

Formula: C₅H₁₂ClNO₂
MW: 153.6100 g/mol
MDL Number: MFCD09910295
CAS Number: 1212377-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR