D-Asparagine monohydrate ≥95%

Supplier: Matrix Scientific

Synonyms: (R)-2-Aminosuccinamic acid hydrate, D-2-Aminosuccinamic acid, H-D-Asn-OH·H₂O, (R)-2,4-Diamino-4-oxobutanoic acid hydrate

075902-10G 075902-100G
102543-548EA 55.38 USD
102543-548 102543-550
D-Asparagine monohydrate ≥95%
D-Asparagine monohydrate

Hazard: Irritant

Formula: C₄H₁₀N₂O₄
MW: 150.14 g/mol (132.12 anhy)
MDL Number: MFCD00149558
CAS Number: 5794-24-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR