DL-Proline ≥95%
Supplier: Matrix Scientific
Synonyms:
(±)-Proline, H-DL-Pro-OH
075901-10G
075901-100G
102543-544EA
38.46
USD
102543-544
102543-546
DL-Proline ≥95%
DL-Proline
Hazard: Irritant
Formula:
C₅H₉NO₂ MW: 115.13 g/mol |
MDL Number:
MFCD00005250 CAS Number: 609-36-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...