2'-Nitroacetophenone ≥95%

Supplier: Matrix Scientific

Synonyms: o-Nitroacetophenone, 1-(2-Nitrophenyl)ethanone

075900-5G 075900-10G
102543-538EA 15.38 USD
102543-538 102543-540
2'-Nitroacetophenone ≥95%
2'-Nitroacetophenone

Hazard: Irritant/Refrigerate

Formula: C₈H₇NO₃
MW: 165.15 g/mol
Boiling Pt: 158-160°C/16mm
Melting Pt: 25-28°C
Density: 1.24
MDL Number: MFCD00007144
CAS Number: 577-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR