2,4-Dibromotoluene ≥95%

Supplier: Matrix Scientific

075899-5G 075899-100G
102543-532EA 38.46 USD
102543-532 102543-536
2,4-Dibromotoluene ≥95%
2,4-Dibromotoluene

Hazard: Irritant/Store in Freezer

Formula: C₇H₆Br₂
Boiling Pt: 243 °C
Melting Pt: –10 °C
MDL Number: MFCD00052985
CAS Number: 31543-75-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR