4-Bromo-3-fluorophenylacetonitrile ≥95%

Supplier: Matrix Scientific

075897-1G 075897-5G 075897-10G
102543-520EA 76.92 USD
102543-520 102543-522 102543-524
4-Bromo-3-fluorophenylacetonitrile ≥95%
4-Bromo-3-fluorophenylacetonitrile

Hazard: Toxic/Light Sensitive

Formula: C₈H₅BrFN
MW: 214.0300 g/mol
Melting Pt: 46…47 °C
MDL Number: MFCD12756199
CAS Number: 499983-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR