α,α,α-Trifluoro-m-toluonitrile ≥95%

Supplier: Matrix Scientific

075895-10G 075895-100G
102543-510EA 72.31 USD
102543-510 102543-512
α,α,α-Trifluoro-m-toluonitrile ≥95%
α,α,α-Trifluoro-m-toluonitrile

Hazard: Irritant

Formula: C₈H₄F₃N
MDL Number: MFCD00001807
CAS Number: 368-77-4
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR