α,4-Dichlorotoluene ≥95%
Supplier: Matrix Scientific
Synonyms:
1-Chloro-4-(chloromethyl)benzene, 4-Chlorobenzyl chloride
075893-10G
075893-100G
102543-500EA
36.92
USD
102543-500
102543-502
α,4-Dichlorotoluene ≥95%
α,4-Dichlorotoluene
Hazard: Irritant
Formula:
C₇H₆Cl₂ MW: 161.03 g/mol |
MDL Number:
MFCD00000914 CAS Number: 104-83-6 UN: 3427 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...