Pyridazine ≥95%

Supplier: Matrix Scientific

075885-1G 075885-5G 075885-10G
102543-462EA 36.92 USD
102543-462 102543-464 102543-466
Pyridazine ≥95%
Pyridazine

Hazard: Irritant

Formula: C₄H₄N₂
MW: 80.09 g/mol
MDL Number: MFCD00006463
CAS Number: 289-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR