3-Fluorophthalonitrile ≥95%

Supplier: Matrix Scientific

075882-1G 075882-5G 075882-10G
102543-448EA 355.38 USD
102543-448 102543-450 102543-452
3-Fluorophthalonitrile ≥95%
3-Fluorophthalonitrile

Hazard: Irritant

Formula: C8H3FN2
MDL Number: MFCD00052458
CAS Number: 65610-13-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR