(S)-(-)-1,1'-Bi-2-naphthol ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-BINOL, (S)-(-)-1,1'-Binaphthyl-2,2'-Diol, (S)-[1,1'-Binaphthalene]-2,2'-diol

075880-100G 075880-25G
102543-442EA 215.38 USD
102543-442 102664-070
(S)-(-)-1,1'-Bi-2-naphthol ≥95%
(S)-(-)-1,1'-Bi-2-naphthol

Hazard: Irritant

Formula: C₂₀H₁₄O₂
MW: 286.33 g/mol
Melting Pt: 206…211 °C
Density: 1.301
MDL Number: MFCD00004068
CAS Number: 18531-99-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR