3-Ethynyl-4-fluoroaniline ≥95%

Supplier: Matrix Scientific

075879-250MG 075879-1G
102543-434EA 367.05 USD
102543-434 102543-436
3-Ethynyl-4-fluoroaniline ≥95%
3-Ethynyl-4-fluoroaniline

Hazard: Irritant

Formula: C8H6FN
MW: 135.14 g/mol
MDL Number: MFCD09029299
CAS Number: 77123-60-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR