Benzo[b]thiophen-2-amine ≥95%

Supplier: Matrix Scientific

075877-250MG 075877-1G
102543-424EA 453.85 USD
102543-424 102543-426
Benzo[b]thiophen-2-amine ≥95%
Benzo[b]thiophen-2-amine

Hazard: Irritant

Formula: C₈H₇NS
MDL Number: MFCD18072620
CAS Number: 4521-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR