Tenofovir disoproxil fumarate ≥95%
Supplier: Matrix Scientific
Synonyms:
(R)-(((((1-(6-Amino-9H-purin-9-yl)propan-2-yl)oxy)methyl)phosphoryl)bis(oxy))bis(methylene) diisopropyl dicarbonate fumarate
075876-1G
075876-5G
102543-420EA
347.69
USD
102543-420
102543-422
Tenofovir disoproxil fumarate ≥95%
Tenofovir disoproxil fumarate
Hazard: Irritant
Formula:
C₁₉H₃₀N₅O₁₀P·C₄H₄O₄ MW: 635.5 g/mol |
MDL Number:
MFCD08141829 CAS Number: 202138-50-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...