N-(4-Bromo-1-methyl-1H-pyrazol-5-yl)pivalamide ≥95%

Supplier: Matrix Scientific

075875-1G 075875-5G 075875-10G
102543-414EA 284.62 USD
102543-414 102543-416 102543-418
N-(4-Bromo-1-methyl-1H-pyrazol-5-yl)pivalamide ≥95%
N-(4-Bromo-1-methyl-1H-pyrazol-5-yl)pivalamide

Hazard: Irritant

Formula: C₉H₁₄BrN₃O
MW: 260.1300 g/mol
MDL Number: MFCD01763678
CAS Number: 679394-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR