2,3,5,6-Tetrafluoro-4-(methoxymethyl)benzyl-2,2-dimethyl-3-(2-methylprop-1-en-1-yl)cyclopropanecarboxylate ≥95%

Supplier: Matrix Scientific

075874-1G 075874-5G
102543-410EA 824.62 USD
102543-410 102543-412
2,3,5,6-Tetrafluoro-4-(methoxymethyl)benzyl-2,2-dimethyl-3-(2-methylprop-1-en-1-yl)cyclopropanecarboxylate ≥95%
2,3,5,6-Tetrafluoro-4-(methoxymethyl)benzyl-2,2-dimethyl-3-(2-methylprop-1-en-1-yl)cyclopropanecarboxylate

Hazard: Irritant

Formula: C₁₉H₂₂F₄O₃
CAS Number: 271241-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR