cis-2-Phenyl-1,3-dioxan-5-ol ≥95%

Supplier: Matrix Scientific

075873-1G 075873-5G 075873-10G
102543-404EA 143.08 USD
102543-404 102543-406 102543-408
cis-2-Phenyl-1,3-dioxan-5-ol ≥95%
cis-2-Phenyl-1,3-dioxan-5-ol

Hazard: Irritant

Formula: C₁₀H₁₂O₃
MW: 180.2000 g/mol
MDL Number: MFCD00274343
CAS Number: 4141-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR