2-(1-Methylguanidino)ethyl dihydrogen phosphate ≥95%

Supplier: Matrix Scientific

Synonyms: Creatinol phosphate, Aplodan, 2-(1-Methylguanidino)ethyl dihydrogen phosphate

075871-1G 075871-5G
102543-396EA 427.69 USD
102543-396 102543-398
2-(1-Methylguanidino)ethyl dihydrogen phosphate ≥95%
2-(1-Methylguanidino)ethyl dihydrogen phosphate

Hazard: Irritant

Formula: C₄H₁₂N₃O₄P
MW: 197.1295 g/mol
MDL Number: MFCD00868297
CAS Number: 6903-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR