Ammonium benzenesulfonate ≥95%

Supplier: Matrix Scientific

075870-1G 075870-5G
102543-392EA 178.46 USD
102543-392 102543-394
Ammonium benzenesulfonate ≥95%
Ammonium benzenesulfonate

Hazard: Irritant

Formula: C₆H₉NO₃S
MW: 175.2100 g/mol
MDL Number: MFCD00191294
CAS Number: 19402-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR