8-(4-Amino-1-methylbutylamino)-6-methoxyquinoline ≥95%

Supplier: Matrix Scientific

Synonyms: Primaquine

075869-1G 075869-5G
102543-388EA 267.69 USD
102543-388 102543-390
8-(4-Amino-1-methylbutylamino)-6-methoxyquinoline ≥95%
8-(4-Amino-1-methylbutylamino)-6-methoxyquinoline

Hazard: Irritant

Formula: C₁₅H₂₁N₃O
MW: 259.3467 g/mol
MDL Number: MFCD00598906
CAS Number: 90-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR