4-Bromo-3-fluoropyridine hydrochloride ≥95%

Supplier: Matrix Scientific

075865-1G 075865-5G 075865-10G
102543-368EA 320 USD
102543-368 102543-370 102543-372
4-Bromo-3-fluoropyridine hydrochloride ≥95%
4-Bromo-3-fluoropyridine hydrochloride

Hazard: Irritant

Formula: C₅H₄BrClFN
CAS Number: 1159811-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR