4-(Piperidin-4-yloxy)pyridine dihydrochloride ≥95%

Supplier: Matrix Scientific

075864-250MG 075864-1G
102543-364EA 826.15 USD
102543-364 102543-366
4-(Piperidin-4-yloxy)pyridine dihydrochloride ≥95%
4-(Piperidin-4-yloxy)pyridine dihydrochloride

Hazard: Irritant

Formula: C₁₀H₁₆Cl₂N₂O
CAS Number: 308386-36-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR