3-Fluoro-5-methoxyphenol ≥95%
Supplier: Matrix Scientific
075863-250MG
075863-1G
075863-5G
102543-358EA
355.38
USD
102543-358
102543-360
102543-362
3-Fluoro-5-methoxyphenol ≥95%
3-Fluoro-5-methoxyphenol
Hazard: Irritant
Formula:
C₇H₇FO₂ MW: 142.1300 g/mol |
MDL Number:
MFCD11848782 CAS Number: 850793-25-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...