3-Amino-1-benzhydrylazetidine mesylate ≥95%

Supplier: Matrix Scientific

075862-1G 075862-5G
102543-354EA 427.69 USD
102543-354 102543-356
3-Amino-1-benzhydrylazetidine mesylate ≥95%
3-Amino-1-benzhydrylazetidine mesylate

Hazard: Irritant

Formula: C₁₇H₂₂N₂O₃S
MDL Number: MFCD20489148
CAS Number: 1373253-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR