(S)-Piperidine-2-carboxylic acid hydrochloride ≥95%

Supplier: Matrix Scientific

075861-1G 075861-5G
102543-350EA 783.08 USD
102543-350 102543-352
(S)-Piperidine-2-carboxylic acid hydrochloride ≥95%
(S)-Piperidine-2-carboxylic acid hydrochloride

Hazard: Irritant

Formula: C₆H₁₂ClNO₂
MW: 165.6200 g/mol
MDL Number: MFCD00066139
CAS Number: 2133-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR