(S)-2-Amino-3-methoxypropanoic acid hydrochloride ≥95%

Supplier: Matrix Scientific

075860-250MG 075860-1G 075860-5G
102543-344EA 2149.23 USD
102543-344 102543-346 102543-348
(S)-2-Amino-3-methoxypropanoic acid hydrochloride ≥95%
(S)-2-Amino-3-methoxypropanoic acid hydrochloride

Hazard: Irritant

Formula: C₄H₁₀ClNO₃
MW: 155.58 g/mol
MDL Number: MFCD03840366
CAS Number: 336100-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR