(4R,5R)-3-tert-Butyl-4-methyl-2,2,5-trimethyloxazolidine-3,4-dicarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: (4R,5R)-3-tert-Butyl 4-methyl 2,2,5-trimethyloxazolidine-3,4-dicarboxylate

075859-1G 075859-5G
102543-340EA 783.08 USD
102543-340 102543-342
(4R,5R)-3-tert-Butyl-4-methyl-2,2,5-trimethyloxazolidine-3,4-dicarboxylate ≥95%
(4R,5R)-3-tert-Butyl-4-methyl-2,2,5-trimethyloxazolidine-3,4-dicarboxylate

Hazard: Irritant

Formula: C₁₃H₂₃NO₅
MW: 273.325 g/mol
MDL Number: MFCD20488086
CAS Number: 1393440-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR