2,3,4-Trifluorotoluene ≥95%

Supplier: Matrix Scientific

075849-1G 075849-5G
102543-298EA 146.15 USD
102543-298 102543-300
2,3,4-Trifluorotoluene ≥95%
2,3,4-Trifluorotoluene

Hazard: Flammable

Formula: C₇H₅F₃
MW: 146.1098 g/mol
Boiling Pt: 126-127°C
Density: 1.222
Flash Pt: 28°C
MDL Number: MFCD00075194
CAS Number: 193533-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR