Methyl-2-bromo-5-nitrobenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Bromobenzoic acid methyl ester, Methyl 2-bromo-5-nitrobenzoate

075847-10G 075847-100G
102543-288EA 143.08 USD
102543-288 102543-290
Methyl-2-bromo-5-nitrobenzoate ≥95%
Methyl-2-bromo-5-nitrobenzoate

Hazard: Irritant

Formula: C₈H₆BrNO₄
MDL Number: MFCD00010867
CAS Number: 6942-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR