1-(5-Bromopyridin-3-yl)-N-methylmethanamine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Bromo-5-[(methylamino)methyl]pyridine

075846-1G 075846-5G
102543-284EA 515.38 USD
102543-284 102543-286
1-(5-Bromopyridin-3-yl)-N-methylmethanamine ≥95%
1-(5-Bromopyridin-3-yl)-N-methylmethanamine

Hazard: Irritant

Formula: C₇H₉BrN₂
MW: 201.06 g/mol
MDL Number: MFCD07375057
CAS Number: 73335-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR