Methyl-3-fluoro-2-nitrobenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-fluoro-2-nitrobenzoate

075845-1G 075845-5G
102543-280EA 235.38 USD
102543-280 102543-282
Methyl-3-fluoro-2-nitrobenzoate ≥95%
Methyl 3-fluoro-2-nitrobenzoate

Hazard: Irritant

Formula: C₈H₆FNO₄
MW: 199.1360 g/mol
MDL Number: MFCD11520393
CAS Number: 1214353-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR