2,3,4,6-Tetra-O-benzyl-D-glucono-1,5-lactone ≥95%

Supplier: Matrix Scientific

075844-1G 075844-5G
102543-276EA 533.85 USD
102543-276 102543-278
2,3,4,6-Tetra-O-benzyl-D-glucono-1,5-lactone ≥95%
2,3,4,6-Tetra-O-benzyl-D-glucono-1,5-lactone

Hazard: Irritant

Formula: C₃₄H₃₄O₆
MW: 538.6302 g/mol
MDL Number: MFCD08703228
CAS Number: 13096-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR