Methyl-2-amino-3-bromobenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-amino-3-bromobenzoate

075843-1G 075843-5G
102543-272EA 75.38 USD
102543-272 102543-274
Methyl-2-amino-3-bromobenzoate ≥95%
Methyl 2-amino-3-bromobenzoate

Hazard: Irritant

Formula: C₈H₈BrNO₂
MW: 230.0586 g/mol
MDL Number: MFCD11042862
CAS Number: 104670-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR