6-Chloro-3-methylpyrimidin-4(3H)-one ≥95%

Supplier: Matrix Scientific

075840-250MG 075840-1G
102543-258EA 684.62 USD
102543-258 102543-260
6-Chloro-3-methylpyrimidin-4(3H)-one ≥95%
6-Chloro-3-methylpyrimidin-4(3H)-one

Hazard: Irritant

Formula: C₅H₅ClN₂O
MW: 144.5600 g/mol
MDL Number: MFCD00955650
CAS Number: 101079-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR