Ethyl-2-((2-ethoxy-2-oxoethyl)amino)-2-oxoacetate ≥95%

Supplier: Matrix Scientific

075839-1G 075839-5G
102543-254EA 427.69 USD
102543-254 102543-256
Ethyl-2-((2-ethoxy-2-oxoethyl)amino)-2-oxoacetate ≥95%
Ethyl-2-((2-ethoxy-2-oxoethyl)amino)-2-oxoacetate

Hazard: Irritant

Formula: C₈H₁₃NO₅
CAS Number: 29655-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR