p-Toluenesulfonylsemicarbazide ≥90.0%

Supplier: Matrix Scientific

075838-10G 075838-100G
102543-250EA 90.77 USD
102543-250 102543-252
p-Toluenesulfonylsemicarbazide ≥90.0%
p-Toluenesulfonylsemicarbazide

Hazard: Irritant

Formula: C8H11N3O3S
MW: 229.26 g/mol
MDL Number: MFCD00072243
CAS Number: 10396-10-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR