4-Bromo-3-picoline hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Bromo-3-methylpyridine hydrochloride

075837-1G 075837-5G 075837-10G
102543-244EA 230.77 USD
102543-244 102543-246 102543-248
4-Bromo-3-picoline hydrochloride ≥95%
4-Bromo-3-picoline hydrochloride

Hazard: Irritant

Formula: C₆H₇BrClN
MW: 208.483 g/mol
MDL Number: MFCD04966778
CAS Number: 40899-37-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR