5-Bromo-2-cyano-4-methylpyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-4-methylpicolinonitrile, 5-Bromo-4-methylpyridine-2-carbonitrile

075831-250MG 075831-1G
102543-218EA 135.38 USD
102543-218 102543-220
5-Bromo-2-cyano-4-methylpyridine ≥95%
5-Bromo-2-cyano-4-methylpyridine

Hazard: Irritant

Formula: C₇H₅BrN₂
MW: 197.0300 g/mol
MDL Number: MFCD07375090
CAS Number: 886364-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR