2-Bromo-3'-(trifluoromethoxy)acetophenone ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-3'-trifluoromethoxyacetophenone, 3-(Trifluoromethoxy)phenacyl bromide

075827-1G 075827-5G 075827-10G
102543-198EA 153.85 USD
102543-198 102543-200 102543-202
2-Bromo-3'-(trifluoromethoxy)acetophenone ≥95%
2-Bromo-3'-(trifluoromethoxy)acetophenone

Hazard: Corrosive/Lachrymator

Formula: C₉H₆BrF₃O₂
MW: 283.0400 g/mol
Boiling Pt: 86 °C
MDL Number: MFCD03094425
CAS Number: 237386-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR