5-(3-Methoxyphenyl)-2H-pyrazol-3-ylamine ≥95%

Supplier: Matrix Scientific

075822-1G 075822-5G
102543-176EA 533.85 USD
102543-176 102543-178
5-(3-Methoxyphenyl)-2H-pyrazol-3-ylamine ≥95%
5-(3-Methoxyphenyl)-2H-pyrazol-3-ylamine

Hazard: Irritant

Formula: C₁₀H₁₁N₃O
MDL Number: MFCD00122783
CAS Number: 96799-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR