4-(Fmoc-amino)-4-piperidinecarboxylic acid hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(Fmoc-Amino)-4-piperidinecarboxylic acid hydrochloride

075821-250MG 075821-1G 075821-5G
102543-170EA 400 USD
102543-170 102543-172 102543-174
4-(Fmoc-amino)-4-piperidinecarboxylic acid hydrochloride ≥95%
4-(Fmoc-amino)-4-piperidinecarboxylic acid hydrochloride

Hazard: Irritant

Formula: C₂₁H₂₃ClN₂O₄
MW: 402.88 g/mol
MDL Number: MFCD03840417
CAS Number: 368866-09-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR