(R)-β-(4-Chlorophenyl)alaninol ≥95%

Supplier: Matrix Scientific

075819-1G 075819-5G
102543-162EA 640 USD
102543-162 102543-164
(R)-β-(4-Chlorophenyl)alaninol ≥95%
(R)-β-(4-Chlorophenyl)alaninol

Hazard: Irritant

Formula: C₉H₁₂ClNO
MDL Number: MFCD07784070
CAS Number: 68208-26-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR