2-Methyl-1,3-cyclopentanedione ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Methylcyclopentane-1,3-dione

075817-5G 075817-10G 075817-100G
102543-152EA 107.69 USD
102543-152 102543-154 102543-156
2-Methyl-1,3-cyclopentanedione ≥95%
2-Methyl-1,3-cyclopentanedione

Hazard: Irritant

Formula: C₆H₈O₂
MW: 112.13 g/mol
MDL Number: MFCD00001406
CAS Number: 765-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR